Industrial Funnels, with short stem
Katalog # SEMA3293
Leverantör: SEMADENI
Specifikationer
- Tratt, y.d.:470 mm
- Förp.:1
- Rör, y.d.:70 mm
Specifikationer
Om denna produkt
HDPE, red
- Robust and strong
- Suspension eye for easy storage
- Good chemical resistance
For filling containers with large quantities of liquid or powdered substances quickly.