Centrifuge tubes, ASTM, cylindrical, conical bottom
Catalog # HECH40930010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Stängningsstorlek:-
- Kapacitet:100 ml
- Förp.:1
Specifications
About this item
Borosilicate glass 3.3, graduated.
- Withstands forces up to 700×g
- Design, accuracy, etc. to meet ASTM D 96 and DIN 51793 specifications