Metallurgical microscopes, AE2000MET
Supplier: MOTIC
Certificates
About this item
These metallurgical microscopes are designed for inspection and analysis of materials, minerals, die-cast parts and precision moulds.
- CCIS® objectives with long working distance for flat and opaque samples
- High quality glasses and anti-reflective coating ensure superb image quality
- Brightfield objectives can be used without restrictions in optical performance
Suitable for use in industrial and scientific research analysis.
Delivery: Supplied with blue filter, Allen hexagonal key, dust cover and power cord.
Specifications
- Catalog No:
- 630-2561
- 630-2554
- 630-2566
- 630-2548
- 630-2549
- 630-2558
- 630-2595
- 630-2560
- 630-2559
- 630-2596
- 630-2594
- MOTI1101001704421
- 630-2555
- 630-2556
- 630-2547
- 630-2557
- Huvud:
- Siedentopf, 45° inclined, 360° rotating (light split 100:0/20:80)
- Siedentopf, 45° inclined, 360° rotating (light split 100:0/20:80)
- Siedentopf, 45° inclined, 360° rotating (light split 100:0/20:80)
- Okular:
- WF 10×/20 mm med diopter
- WF 10×/20 mm med diopter
- WF 10×/20 mm med diopter
- Revolver:
- 5 lägen, inclined
- 5 lägen, inclined
- 5 lägen, inclined
- Objektbord:
- 280×192 mm (movement: 50×50 mm)
- 280×192 mm (movement: 50×50 mm)
- 280×192 mm (movement: 50×50 mm)
- Fokussystem:
- Koaxial grov- och fininställning
- Koaxial grov- och fininställning
- Koaxial grov- och fininställning
- Belysning:
- Halogen
- Halogen
- Halogen