Waist wader, Steelite™ Safety, FW71
Leverantör: Portwest
Certifikat
Om denna produkt
Heavy-duty PVC/nitrile safety waist wader with adjustable thigh straps and integrated S5 safety boots.
- Colour-coded red outsole for easy identification of safety features
- Slip-and oil resistant, anti-static outsole
- Energy absorber heel
- Toe protection cap made from steel (200 J)
- Pierce resistant steel midsole
Colour: Black
EN ISO 20345:2011, S5
Certifications: EN ISO 20345:2011, S5
Specifikationer
- Catalog No:
- PRTWFW71BKR43
- PRTWFW71BKR37
- PRTWFW71BKR38
- PRTWFW71BKR39
- PRTWFW71BKR44
- PRTWFW71BKR46
- PRTWFW71BKR41
- PRTWFW71BKR40
- PRTWFW71BKR45
- PRTWFW71BKR42
- PRTWFW71BKR47
- Modell:
- FW71
- FW71
- FW71
- FW71
- FW71
- FW71
- FW71
- FW71
- FW71
- FW71
- FW71
- Outfit applikationer:
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Waste water, Industrial
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tyg:
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile
- PVC/nitrile