Hand sanitiser wipes, IW40
Katalog # PRTWIW40BLU
Leverantör: Portwest
Specifikationer
- Förp.:200 per dispenser
- Beskrivning:Hand sanitising wipes, polyviscose, blue
- L×B:200×200 mm
Specifikationer
Om denna produkt
Polyviscose wipes with anti-bacterial and anti-fungal action - effective against 99,9% of bacteria including Human influenza H1N1 and H3N2.
- Practical dispenser box for single sheet dispensing
- For hygienic hands
Certifications: EN 12054, EN 14476
Caution: Use biocides safely. Always read the label and product information before use.