Wipes for hands cleaning, IW10
Catalog # PRTWIW10WHR
Supplier: Portwest
Specifications
- Förp.:150 per dispenser
- Färg:Vit
- L×B:260×270 mm
Specifications
About this item
These PP wipes remove wet paint, oil, grease and grime. For use on hands, tools and surfaces.
- Antibacterial action: Effective against E-coli, Salmonella, MRSA, Listeria and Weils Disease
- Dermatologically tested
- Packed in practical dispensing bucket
Caution: Use biocides safely. Always read the label and product information before use.