Stand for Imhoff sedimentation cones
Katalog # BEHRB00179003
Leverantör: BEHR
Specifikationer
- Beskrivning:For two cones with stopcock
- Förp.:1
Specifikationer
Om denna produkt
Rack for Sedimention vessels, made of solid PVC grey, for two Imhoff cones with stopcock.