Examination gloves, latex, Mutexil
Supplier: MUTEXIL
Certificates
About this item
These disposable gloves are suitable for use in medical environments, for food processing and in laboratories.
- Non sterile, powder-free, smooth finish
- Supplied in convenient dispenser boxes
Certifications: EN 455; AQL 1,5
Food contact approved
Food contact approved
Specifications
- Catalog No:
- MAKA8883150/S
- MAKA8883150/M
- MAKA8883150/L
- MAKA8883150/XL
- Modell:
- Mutexil
- Mutexil
- Mutexil
- Mutexil
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Smooth
- Smooth
- Smooth
- Smooth
- EN standard:
- EN 455
- EN 455
- EN 455
- EN 455
- Godkända för livsmedelskontakt:
- +
- +
- +
- +