Spoon spatula, bent
Katalog # USBE3317
Leverantör: USBECK Laborgeräte
Specifikationer
- Längd:185 mm
- Förp.:1
- Skedstorlek:19 mm
Specifikationer
Om denna produkt
Double-ended stainless steel spatula with bent spoon end and flat spatula end.
Double-ended stainless steel spatula with bent spoon end and flat spatula end.