Office lamps, SN/SNE
WLDM111581000
: WALDMANN
- Beskrivning:Universal light
- Förp.:1
Light with aluminiumised parabolic louvre, model SNE 118.
- 18 W fluorescent lamp
- Electronic ballast operating device
Weight: 2,8 kg
Light with aluminiumised parabolic louvre, model SNE 118.
Weight: 2,8 kg