Industrial gloves, 377
Leverantör: Showa
Certifikat
Om denna produkt
These seamless polyester/nylon knitted gloves feature a nitrile foam coating which protects the hand from oils, hydrocarbons, grease and abrasion. As the gloves are impermeable they are suitable for working in damp or greasy environments.
- Fully coated, extra coating on palm
- Reinforced knit cuff, scalloped edge, anatomical shape
- Robust, great dexterity and good tearing resistance
- Optimal long lasting grip
Ideal for light assembly tasks in construction industries, in the maritime sector and in petrochemical industries.
Certifications: EN 388:2016 (4121X); Cat. II
Specifikationer
- Catalog No:
- 112-5809
- 112-5812
- 112-5808
- 112-5810
- 112-5811
- Modell:
- 377
- 377
- 377
- 377
- 377
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Yta:
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Fully coated + extra palm coating
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)