Nitrile gloves, Dermatril® P 743
These disposable, chemical resistant gloves provide a high level of protection against chemical and bacteriological contamination. They are also resistant against cytotoxic agents and viruses.
- Free from latex and silicone
- Very good tactual sensation, excellent wearing comfort
- Increased chemical and mechanical resistance compared to standard Dermatril® gloves
- Improved grip thanks to roughened fingertips
- Long cuff provides protection for the forearm
Ideal for use in laboratory operations, in chemical and electronic industries, as well as in food processing.
Length: 270 - 290 mm
Thickness: 0,2 mm
Colour: Blue
EN 374 (JKL), EN 455-1/-2/-3, Cat. III, AQL 0,65 Food contact approved
: EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455, Cat. III, AQL 0,65
Food contact approved
Food contact approved
- Catalog No:
- 112-4313
- 112-4315
- 112-4314
- 112-4316
- 112-4317
- 112-4318
- Modell:
- P 743
- P 743
- P 743
- P 743
- P 743
- P 743
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455
- EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455
- EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455
- EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455
- EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455
- EN 388:2016 (200XX), EN ISO 374-1 Type A (JKLOPT), EN 455
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +
- +