Nitrile gloves, Dermatril® L 741
Leverantör: KCL by Honeywell
Certifikat
Om denna produkt
These disposable, chemical resistant gloves provide a high level of protection against chemical and bacteriological contamination. They are also resistant against cytotoxic agents and viruses.
- Free from latex and silicone
- Very good tactual sensation, excellent wearing comfort
- Increased tear resistance
- Improved grip thanks to roughened fingertips
- Long cuff provides protection for the forearm
Ideal for use in laboratory operations, in chemical and electronic industries, as well as in food processing.
Length: 270 - 280 mm
Thickness: 0,11 ± 0,03 mm
Colour: Blue
EN 374, EN 455-1/-2/-3, Cat. III, AQL 0,65 Food contact approved
Certifications: EN ISO 374-1 Type B (KPT), EN 455, Cat. III, AQL 0,65
Food contact approved
Food contact approved
Specifikationer
- Catalog No:
- 112-1743
- 112-1744
- 112-1741
- 112-1742
- KCHL741.11
- Modell:
- L 741
- L 741
- L 741
- L 741
- L 741
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN ISO 374-1 Type B (KPT), EN 455
- EN ISO 374-1 Type B (KPT), EN 455
- EN ISO 374-1 Type B (KPT), EN 455
- EN ISO 374-1 Type B (KPT), EN 455
- EN ISO 374-1 Type B (KPT), EN 455
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +