Cytological standard stain acc. to Szczepanik, CYTOCOLOR®, Sigma-Aldrich®
Catalog # 1.15355.0001
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Fibre carton
- Förp.:6
- Seq:0006
- Pk Info:Preparation Capacity 1000 slides
Specifications
About this item
:
UN:1219
ADR:3,
:GHS02
:GHS08
:GHS07
Storage Temperature:Omgivande
:1