Pliers for stainless steel tubing
Catalog # 554-1105
Supplier: VICI JOUR RESEARCH
Specifications
- Beskrivning:Tubing pliers, for cutting 1/16" stainless steel tubing off a coil
- Förp.:1
Specifications
About this item
The pliers type of cutter can be used for quick cuts for non-critical applications or at hard to get places.
- For easy cutting of ¹/₁₆" tubing off a coil