Nitrile gloves, Dermatril® 740
Leverantör: KCL by Honeywell
Certifikat
Om denna produkt
These disposable nitrile gloves offer a high level of protection against chemical and bacterial contamination as well as impressive mechanical resistance.
- Very good tactile sensitivity and outstanding comfort thanks to the smooth fit
- Increased tear resistance and improved grip due to textured fingertips
- With beaded cuffs
- Free from silicone and allergens
- Resistant to a variety of cytostatics and resistant to viruses in accordance with ASTM F 1671:2007
- Hygienic dispenser for individual dispensing
Ideal for use in the chemical, food, electronics and computer industries as well as for cleaning and maintenance work.
Length: 240 - 260 mm
Thickness: 0.11 ± 0.03 mm
Colour: blue
EN 374-2; EN 455-1/2, EN 388 (0000); Cat. III; AQL 0.65 Suitable for food use
Certifications: EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS; Cat. III; AQL <0,65
Food contact approved
Food contact approved
Specifikationer
- Catalog No:
- 112-2985
- KCHL740.6
- 112-1733
- 112-1734
- 112-1731
- 112-1732
- Modell:
- 740
- 740
- 740
- 740
- 740
- 740
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Textured fingertips
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS
- EN 455, EN ISO 374-1 Type B (KPT); EN ISO 374-5 VIRUS
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +
- +