Ultrasonic baths, SONOREX DIGITEC
Supplier: BANDELIN-ELECTRONIC
Certificates
About this item
SONOREX DIGITEC is the high-power ultrasonic bath with keypress operating system for aqueous cleaning solutions.
- Heating from 20 to 80 °C, thermostatically adjustable, LED
- Optical signal in case of excess temperature
- Automatic short-time ultrasound during heating up period to avoid delay in boiling - switch-off possible
- Timer for countdown or continuous operation
- DEGAS - quick degassing
- SweepTec® - permanent sound field oscillation ensures even and gentle cleaning
- Automatical safety shut-down 12 hours after last keypress
- Marking of filling level for safe dosage
- Drain valve for simple emptying, one-piece welded drain bow
Protection class IP 33 at highest possible level through liquid-tight mains cable and housing front without openings.
Specifications
- Catalog No:
- BAND3220
- BAND3231
- BAND3235
- BAND3266
- BAND3257-GB
- BAND3245
- BAND3240
- Modell:
- DT 31 H
- DT 1028 H
- DT 102 H
- DT 1028 CH
- DT 1028 GB
- DT 510
- DT 255 H
- Volym:
- 0,9 L
- 28,0 L
- 3,0 L
- 45,0 L
- 9,5 L
- 9,7 L
- 5,5 L
- Frekvens:
- 35 kHz
- 35 kHz
- 35 kHz
- 35 kHz
- 35 kHz
- 35 kHz
- 35 kHz
- Uppvärmningseffekt:
- 70 W
- 1300 W
- 140 W
- 1450 W
- -
- 280 W
- Max. uteffekt:
- 240 W
- 1200 W
- 480 W
- 1200 W
- 1280 W
- 640 W
- 640 W
- Innermått, B×D×H:
- 190×85×60 mm
- 500×300×200 mm
- 240×140×100 mm
- 500×300×300 mm
- 500×300×65 mm
- 300×240×150 mm
- 300×150×150 mm
- W×D×H:
- 205×100×170 mm
- 535×325×400 mm
- 260×160×250 mm
- 540×340×500 mm
- 535×325×205 mm
- 325×265×305 mm
- 325×175×295 mm
- Vikt:
- 1,9 kg
- 14,7 kg
- 4,3 kg
- 23,7 kg
- 7,0 kg
- 5,3 kg