Mini-Torso, without head, 9-part
Mini-Torso, without head, 9-part
Catalog # BINH1000194
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
This human mini torso is approximately half life-size, supplied with the 3B Torso Guide. Even small hands can quickly disassemble and assemble this torso. This torso includes the following parts and organs:
- Two lungs
- Two-part heart
- Stomach
- Liver with gall bladder
- Two-part intestinal tract
Dimensions: 42×24×18 cm
Weight: 1,59 kg
Specifications
- Beskrivning:Mini-Torso, without head, 9-part