Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Belysning
UV-lampor
Hand-held UV light
Hand-held UV light
Hand-held UV light
KOEHE-1450
:
KOEHLER TECHNISCHE PRODUKTEN
Hand-held UV light
KOEHE-1450
:
KOEHLER TECHNISCHE PRODUKTEN
Beskrivning:
UV lamp with robust powder-coated metal housing, 45×145 mm
Effekt:
6 W
Förp.:
1
Våglängd:
254/366 nm