Immersion probe
Immersion probe, NiCr-Ni, class 1, flexible jacket material (inconel), electrically isolated, −200…+1150 °C, 1,5×130 mm
Katalog # 620-4707
Leverantör: Greisinger
Gå till:
Product Details & Documents
Specifikationer
- Mätområde:−200…+1150 °C
- Beskrivning:Immersion probe, NiCr-Ni, class 1, flexible jacket material (inconel), electrically isolated
- Ø×L:1,5×130 mm