Temperature probe
Katalog # 620-4728
Leverantör: Greisinger
Specifikationer
- Mätområde:−50…+400 °C
- Beskrivning:Immersion probe, PT 100, class B, non-corrosive stainless steel with plastic handle and one-meter four-wire PVC cable
- Förp.:1
- Ø×L:3×150 mm