Adhesion slides, Apex Superior
Catalog # 630-2184
Supplier: LEICA HISTOLOGY
Specifications
- B×D:76×26 mm
- Färg:Vit
- Beskrivning:Adhesion slides
- Förp.:72
Specifications
About this item
These adhesive slides are coated for a positive charge to enhance hydrophilicity.
- Homogeneous covering by aqueous stains and reagents
- Consistent full slide staining
For demanding procedures such as standard IHC and HIER techniques.