ContactUsLinkComponent
Hydrometers, Anti-freeze Tester
Hydrometers, Anti-freeze Tester
Catalog # AMARH851710
Supplier:  Amarell
Hydrometers, Anti-freeze Tester
Catalog # AMARH851710
Supplier:  Amarell
Specifications
  • Längd:
    370/25 mm
  • Förp.:
    1
  • Mätområde:
    −30…0 °C
Specifications
About this item
In this hydrometer in glass tube with suction ball and plastic suction.
  • Paper scale with coloured indication the charge of indication °C status
  • Shot weighted body
  • Complete in cardboard box or in blister packing