Beauchene adult human skull model
Katalog # BINH1000069
Leverantör: 3B Scientific
Specifikationer
- Vikt:0,7 kg
- Beskrivning:Beauchene adult human skull didactic coloured version in 22 parts
- B×D×H:210×140×160 mm
- Förp.:1
Specifikationer
Om denna produkt
Beauchene adult human skull is a natural cast and makes the complexity of the anatomical structure easy to understand, since it is disassembled into its 22 individual bones.
- Bones/bone pairs are coloured to make dentification easier
- Bones can be reassembled by means of inconspicuous, stable connectors attached at the slightly simplified skull sutures
- Human skull consists of individual bones that gradually grow together as the development proceeds
This skull model is a great tool for teaching and learning the complex anatomy of the human skull.