Ultrasonic baths, Bransonic®
Leverantör: BRANSON
Certifikat
Om denna produkt
M and MH Series ultrasonic baths are known for offering a high level of precision and reliability.
- Sweep frequency eliminates standing waves and creates consistent cavitation throughout the tank
- 40 kHz rugged industrial transducers
- Controls are located above and behind the cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions
- Plastic body, chemical-resistant
M Series includes 60-min mechanical timer and continuous operation.
MH Series includes 60-min mechanical timer and continuous operation with heating capability.
Specifikationer
- Catalog No:
- 142-0136
- 142-0137
- 142-0128
- 142-0133
- 142-0139
- 142-0130
- 142-0131
- 142-0140
- 142-0127
- 142-0134
- Modell:
- M5800-E
- M5800H-E
- M1800H-E
- M3800-E
- M8800-E
- M2800-E
- M2800H-E
- M8800H-E
- M1800-E
- M3800H-E
- Volym:
- 9,5 L
- 9,5 L
- 1,9 L
- 5,7 L
- 20,8 L
- 2,8 L
- 2,8 L
- 20,8 L
- 1,9 L
- 5,7 L
- Frekvens:
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- 40 kHz
- Innermått, B×D×H:
- 290×240×150 mm
- 290×240×150 mm
- 150×140×100 mm
- 290×150×150 mm
- 495×290×150 mm
- 230×140×100 mm
- 230×140×100 mm
- 490×290×150 mm
- 150×140×100 mm
- 290×150×150 mm
- W×D×H:
- 397×401×378 mm
- 397×401×378 mm
- 251×304×302 mm
- 397×317×377 mm
- 596×466×391 mm
- 337×304×302 mm
- 337×304×302 mm
- 596×466×391 mm
- 251×304×302 mm
- 397×317×377 mm
- Vikt:
- 6,4 kg
- 6,4 kg
- 3,2 kg
- 5,4 kg
- 11,8 kg
- 4,1 kg
- 4,1 kg
- 11,8 kg
- 3,2 kg
- 5,4 kg