Double spatulas, Chattaway type
Katalog # HAMMHSN193-18
Leverantör: HAMMACHER KARL
Specifikationer
- Längd:180 mm
- Bladbredd:8 mm
- Förp.:1
Specifikationer
Om denna produkt
Double-ended Chattaway spatulas made of stainless steel. With one bent end.