ContactUsLinkComponent
Hand cleansers, Estesol® premium sensitive
Hand cleansers, Estesol® premium sensitive
Katalog # EVON32013
Leverantör:  Deb-STOKO
undefined
Hand cleansers, Estesol® premium sensitive
Katalog # EVON32013
Leverantör:  Deb-STOKO
Specifikationer
  • Beskrivning:
    Hand cleanser, Estesol® premium sensitive, perfume-free, in 2000 ml soft bottle for Stoko Vario® dispensers
  • Seq:
    02
  • Förp.:
    6
Specifikationer
Om denna produkt
Estesol® premium sensitive is a non soapy, liquid skin cleanser particularly suitable for gently cleansing of heavily stressed skin that has already suffered damage. For Light to medium-duty skin cleansing. Suitable for the whole body.
  • Contains natural moisturisers
  • Skin friendly pH
  • Perfume-free, and free from preservatives, colourants and silicone
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904