Single channel pipettes, mechanical, BagPipet®
Catalog # 432-0012
Supplier: INTERSCIENCE
Specifications
- Volym:0,1/0,9/1 ml
- Beskrivning:Autoclavable pipette, for use with BagTips®
- Förp.:1
Specifications
About this item
BagPipet® is an autoclavable pipette with three selectable volumes (0,1 ml / 0,9 ml / 1 ml). It is used with BagTips®, a single-use sterile and rigid straw.
- Economical
- No risk of cross-contamination
- Light and ergonomic
- Autoclavable and calibrated according to the GLP