Magnetic stirring bars, PTFE, micro
Catalog # COWI001.830
Supplier: COWIE
Specifications
- Ø:3 mm
- Typ:Micro, cylindrical
- Längd:30 mm
- Förp.:1
Specifications
About this item
Magnetic stirrer bar with white PTFE coating, used for very small containers.