Critical care syringes, BD Vacutainer®
Katalog # BDAM364356
Leverantör: BD Medical
Specifikationer
- Kapacitet:1,0 ml
- Beskrivning:BD A-Line™ critical care syringes, without needle, slip tip, with 30 units of spray dried heparin
- Förp.:100
Specifikationer
Om denna produkt
Critical care syringes can be used to measure arterial blood gas, ionised calcium, ionised magnesium, as well as perform critical care tests.
- Permits aspiration
Syringes contain calcium balanced lithium heparin spray dried on the sides of the syringe.