Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Sudan II
Sudan II ≥95.0% (by UV/Vis)
Sudan II ≥95.0% (by UV/Vis)
Sudan II ≥95.0% (by UV/Vis)
TCIAO0101-25G
:
TCI
:
Sudan II ≥95.0% (by UV/Vis)
TCIAO0101-25G
:
TCI
:
Appearance:
Orange to Red powder to crystal
Purity (UV-vis method):
min. 95.0 %
Melting point:
158.0 to 163.0 deg-C
Lambda max.:
496.0 to 500.0 nm (CHCl3)