2-(Allylthio)-2-thiazoline för syntes, Sigma-Aldrich®
Catalog # 8.18083.0010
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:10 ml
- Seq:2275
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Molar Mass:159.28 g/mol
- Boiling Point:92 - 94 °C (2 hPa)
- Density:1.16 g/cm3 (20 °C)
Specification Test Results
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:222-679-4
UN:2810
:GHS06
Storage Temperature:Omgivande
CAS Number:3571-74-2
Formula:C₄H₇NS