Racks for Leighton tissue culture tubes
Catalog # BELC1917-00016
Supplier: BELLCO GLASS
Specifications
- för rör:125 - 150 mm
- Ant. hål:36
- Beskrivning:Self locking, for Leighton type tissue culture tubes
- Förp.:1
Specifications
About this item
Stainless steel.
- Spring clip prevents tube from moving in rack
- Excellent chemical resistance
- Excellent temperature resistance
For Leighton type culture tubes.