VWR® Acrylamide-Bis Solution (37,5:1) 30% Aqueous Solution, Ultra Pure Grade
Katalog # E347-500ML
Leverantör: VWR Chemicals
Specifikationer
- Beskrivning:Acrylamide-Bis solution (37,5:1) 30% aqueous solution
- Förpackningstyp:Plastic bottle
- Förp.:500 mL