VWR® YPD, Broth, for Biotechnology
Catalog # J903-500G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle for solids
- Förp.:500 g
- Beskrivning:YPD broth
Specifications
About this item
Yeast peptide dextrose (YPD) broth is used for the propagation and maintenance of yeasts.
YPD broth is supplied as a pre-mixed powder which, when dissolved at 50 g/l of water, yields peptone (20 g/l), yeast extract (10 g/l), glucose (20 g/l).
Final pH of solution (25 °C): 6,5 ±0.5
Autoclave or sterile filter.