Metylrött 0,04% i vatten for pH determination
TCIAM0056-500ML
: TCI
:
- Förpackningstyp:Glass bottle
- Förp.:500 mL
- Seq:2725
- Appearance:Light yellow to Yellow to Orange clear liquid
- Transition interval (pH):(red)4.2 to 6.2 (yellow)
:
:14,06119
:269.3 g/mol
:207-776-1
:493-52-7
:C₁₅H₁₅N₃O₂
:MFCD00002425