Order Entry
Sweden
ContactUsLinkComponent
Salpetersyra 0.05 mol/l (0.05 N)
Salpetersyra 0.05 mol/l (0.05 N)
Catalog # FFCHFF62919021
Supplier:  FF CHEMICALS
CAS Number:  
Salpetersyra 0.05 mol/l (0.05 N)
Catalog # FFCHFF62919021
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Förp.:
    20 L
  • Seq:
    5302

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Omgivande
EINECS:231-714-2
UN:3264
ADR:8,III
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349