Functional model of the hand and finger joints
Catalog # SOMMNS55
Supplier: SOMMER MARCUS
Specifications
- Vikt:0,4 kg
- Beskrivning:Functional model of the hand and fingers
- Material:SOMSO-Plast®
- B×D×H:360×180×190 mm
- Förp.:1
Specifications
About this item
The following movements are possible:
- Flexion and extension
- Abduction and adduction
- Opposition and reposition of the thumb
- Dorsal and palmar flexion
- Radial abduction
- Ulnar abduction of the hand
Packaging: Removable from stand with green base.