Functional model of the hip joint
Catalog # SOMMNS51
Supplier: SOMMER MARCUS
Specifications
- Vikt:1,25 kg
- Beskrivning:Functional model of the hip
- Material:SOMSO-Plast®
- B×D×H:350×200×180 mm
- Förp.:1
Specifications
About this item
The following movements are possible:
- Raising of the thigh, anteversion
- Retracting of the thigh, retroversion
- Lifting up of the thigh to the side, abduction
- Inner and outer rotation
Delivery: Removable from stand with green base.