Functional model of the knee
Catalog # SOMMNS50
Supplier: SOMMER MARCUS
Specifications
- Vikt:1 kg
- Beskrivning:Functional model of the knee joint
- Material:SOMSO-Plast®
- B×D×H:340×180×180 mm
- Förp.:1
Specifications
About this item
The following movements are possible:
- Flexion
- Extension
- Inner and outer rotation