Section through the shoulder joint
Katalog # SOMMNS48
Leverantör: SOMMER MARCUS
Specifikationer
- Beskrivning:Section through the shoulder joint
- Förp.:1
Specifikationer
Om denna produkt
Frontal section. Model in one piece.
- Natural size
- Made in SOMSO-Plast®
H×W×D: 26×32×4 cm
Weight: 0,9 kg