Section through a normal foot
Section through a normal foot
Katalog # SOMMNS47
Leverantör: SOMMER MARCUS
Gå till:
Product Details & Documents
Sagittal section. Model in one piece.
- Natural size
- Made in SOMSO-Plast®
H×W×D: 26×32×4 cm
Weight: 0,8 kg
Specifikationer
- Beskrivning:Section through a normal foot