Sections of the brain
Katalog # SOMMBS45
Leverantör: SOMMER MARCUS
Specifikationer
- Vikt:5,1 kg
- Beskrivning:Sections of the brain (5)
- B×D×H:570×100×490 mm
- Förp.:1
Specifikationer
Om denna produkt
Made in SOMSO-Plast® and with the following features:
- 1. Left hemisphere - Cortical relief of the cerebrum and cerebellum
- 2. Median section through the brain
- 3. Cerebral ventricle (opened) and stem ganglia (viewed from above)
- 4. Horizontal section through the left hemisphere. Right: Lateral ventricle (opened) and adjoining nuclei with choroid plexus (cf. No. 3)
- 5. Frontal section through the brain and brain stem with the subcortical nuclei and projection tracts
Packaging: In display case with transparent cover.