Section through the hip joint
Section through the hip joint
Katalog # SOMMNS44
Leverantör: SOMMER MARCUS
Gå till:
Product Details & Documents
Frontal section. Model in one piece.
- Natural size
- Made in SOMSO-Plast®
H×W×D: 26×32×4 cm
Weight: 0,9 kg
Specifikationer
- Beskrivning:Section through the hip joint