Section through the hand
Section through the hand
Katalog # SOMMNS45
Leverantör: SOMMER MARCUS
Gå till:
Product Details & Documents
Sagittal section. Model in one piece.
- Natural size
- Made in SOMSO-Plast®
H×W×D: 26×32×4 cm
Weight: 0,8 kg
Specifikationer
- Beskrivning:Section through the hand