Cable for CCD sensor
Cable for CCD sensor
Katalog # DIDAPOF010610
Leverantör: DIDALAB
Gå till:
Produktdetaljer och dokument
This cable retrieves the “raw” signals of the sens interface. It helps to explain the principle of CCD photosensitive sensors and show the relation between the light received by the sensor and the electrical measure.
Specifikationer
- Beskrivning:Cable for CCD sensor