CCD camera, POF 010 300
Katalog # DIDAPOF010300
Leverantör: DIDALAB
Specifikationer
- Beskrivning:Basic CCD camera
- Förp.:1
Specifikationer
Om denna produkt
This Caliens camera enables measuring very precise distances.
- CCD sensor, 2048 pixels
- User-friendly software, Windows compatible
- Power supplied via USB port (connected to a PC)
Delivery: Supplied with Ø 10 mm rod for use with optical benches, software and USB cable.