Sealing film, self-adhesive for ELISA, PCR
Katalog # 211-0001
Leverantör: Brand
Specifikationer
- Optisk egenskap:Transparent
- Beskrivning:Sealing film, self-adhesive, PP, for PCR and ELISA, single sheets
- Seq:01
- Förp.:100
Specifikationer
Om denna produkt
Transparent PP film allows the visual inspection of the samples. Temperature range: −40 to +125 °C.