High visibility rain trousers, Norvill 799Z
: SIOEN
Water- and windproof trousers made from highly breathable Siopor® Ultra fabric consisting of 100% polyester fabric (plain weave) with 100% PU coating, with fixed polyamide lining.
- Taped seams on the inside
- Legs adjustable by press studs
- Elasticated waist
- High tear resistance
The fabric is water-repellent on the outside and features a moisture attracting coating on the inside in order to keep the body warm and dry.
Weight: 155 g/m²
: EN 471 (1-2), EN 343 (3-3)
- Catalog No:
- SIOE799Z/278/1XL
- SIOE799Z/278/L
- SIOE799Z/278/S
- SIOE799Z/278/M
- SIOE799Z/279/M
- SIOE799Z/278/3XL
- SIOE799Z/279/S
- SIOE799Z/279/3XL
- SIOE799Z/279/L
- SIOE799Z/278/2XL
- SIOE799Z/279/2XL
- SIOE799Z/279/1XL
- Modell:
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Norvill 799Z
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Rain (waterproof)
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Tyg:
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- 100% polyester fabric (plain weave) with 100% PU coating
- Vikt:
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²
- 155 g/m²