Fleece lining for jackets, flame-retardant, Watson 7221
: SIOEN
Detachable lining made of double-sided, flame-retardant polyester with anti-pilling treatment. Can be zipped into jackets 7218 & 7219.
- Good cold protection, breathable
- Zip closure
- Slit opening to inside pocket of rain jacket
- Round inset sleeves, knitted windcuffs at end of sleeves
- Can not be worn separately
Weight: 275 g/m²
: EN ISO 14116 (Index 1), EN 14058 (1XXXX)
- Catalog No:
- SIOE7221TF1B75/M
- SIOE7221TF1B75/3XL
- SIOE7221TF1B75/S
- SIOE7221TF1B75/L
- SIOE7221TF1B75/1XL
- SIOE7221TF1B75/2XL
- Modell:
- Watson 7221
- Watson 7221
- Watson 7221
- Watson 7221
- Watson 7221
- Watson 7221
- Outfit applikationer:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Vikt:
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²